accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COOH3COOCH3

CAS RN 944392-68-1

IUPAC Name: 1,3-dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate
Molecular Weight (g/mol): 320.15
Molecular Formula: C16H21BO6
InChi Key: IGSNWXAGFXHYOG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)B1OC(C)(C)C(C)(C)O1)C(=O)OC
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)