accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2BrFFF

CAS RN 944401-56-3

IUPAC Name: 5-bromo-4-(trifluoromethyl)pyridin-2-amine
Synonyms: 5-bromo-4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 241.01
Molecular Formula: C6H4BrF3N2
InChi Key: AEWYLVDLWQPJGW-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)