accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944401-77-8

IUPAC Name: 4-fluoropyridin-2-amine
Synonyms: 4-fluoropyridin-2-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: WCCFLOQQACDOAX-UHFFFAOYSA-N
SMILES: NC1=NC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results