accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3Pt

CAS RN 94442-22-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 319.31
Molecular Formula: C9H16Pt
InChi Key: —
SMILES: C[Pt](C)C.Cc1cccc1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)