accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOBrHBr

CAS RN 944450-78-6

IUPAC Name: 2-bromo-1-(1-methyl-1H-1,3-benzodiazol-5-yl)ethan-1-one hydrobromide
Synonyms: 2-bromo-1-(1-methyl-1,3-benzodiazol-5-yl)ethanone hydrobromide
Molecular Weight (g/mol): 334.01
Molecular Formula: C10H10Br2N2O
InChi Key: LRWGKOQDEBXXPS-UHFFFAOYSA-N
SMILES: Br.CN1C=NC2=CC(=CC=C12)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results