accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944450-82-2

IUPAC Name: [(4-bromothiophen-3-yl)methyl](methyl)amine hydrochloride
Synonyms: [(4-bromothiophen-3-yl)methyl](methyl)amine hydrochloride
Molecular Weight (g/mol): 242.56
Molecular Formula: C6H9BrClNS
InChi Key: GORGMQYTNXIOBQ-UHFFFAOYSA-N
SMILES: Cl.CNCC1=CSC=C1Br
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results