accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHOSH2OHCl

CAS RN 944450-84-4

IUPAC Name: 1-[(thiophen-2-yl)methyl]piperidine-4-carboxylic acid hydrate hydrochloride
Synonyms: 1-(thiophen-2-ylmethyl)piperidine-4-carboxylic acid hydrate hydrochloride
Molecular Weight (g/mol): 279.78
Molecular Formula: C11H18ClNO3S
InChi Key: YUUKLZNSOZECHH-UHFFFAOYSA-N
SMILES: O.Cl.OC(=O)C1CCN(CC2=CC=CS2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results