accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944450-95-7

IUPAC Name: 5-[(methylamino)methyl]-N-phenylpyrimidin-2-amine
Synonyms: 5-[(methylamino)methyl]-N-phenylpyrimidin-2-amine
Molecular Weight (g/mol): 214.27
Molecular Formula: C12H14N4
InChi Key: IFWJJBILYKUROD-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(NC2=CC=CC=C2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results