accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNOOHOHCl

CAS RN 944450-97-9

IUPAC Name: 5-[(morpholin-4-yl)methyl]-1,2-oxazole-3-carboxylic acid hydrochloride
Synonyms: 5-(morpholin-4-ylmethyl)-1,2-oxazole-3-carboxylic acid hydrochloride
Molecular Weight (g/mol): 248.66
Molecular Formula: C9H13ClN2O4
InChi Key: OOOBOQOTQIVUQQ-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=NOC(CN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results