accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944451-10-9

IUPAC Name: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane) hydrochloride ruthenium chloride
Synonyms: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane) hydrochloride ruthenium chloride
Molecular Weight (g/mol): 1898.31
Molecular Formula: C106H107Cl5NP4Ru2
InChi Key: LJSDQAUPXVWQNF-UHFFFAOYSA-M
SMILES: Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results