accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944451-32-5

IUPAC Name: (4-{5-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane; 1-methyl-4-(propan-2-yl)benzene; dichlororuthenium
Synonyms: (4-{5-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane; cymene; dichlororuthenium
Molecular Weight (g/mol): 1485.75
Molecular Formula: C84H114Cl2O8P2Ru
InChi Key: MYYXUILRYSITRR-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.COC1=C(C=C(C=C1C(C)(C)C)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results