accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiOOFFFFFOCH3OCH3

CAS RN 944721-52-2

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 11-(trimethoxysilyl)undecanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 11-(trimethoxysilyl)undecanoate
Molecular Weight (g/mol): 472.52
Molecular Formula: C20H29F5O5Si
InChi Key: KSYPNQIBGIOYPZ-UHFFFAOYSA-N
SMILES: CO[Si](CCCCCCCCCCC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results