accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94525-05-0

ONH2FFFOH2NFFF

CAS RN 94525-05-0

IUPAC Name: 4-{4-[4-amino-2-(trifluoromethyl)phenoxy]phenoxy}-3-(trifluoromethyl)aniline
Synonyms: 4-{4-[4-amino-2-(trifluoromethyl)phenoxy]phenoxy}-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 428.33
Molecular Formula: C20H14F6N2O2
InChi Key: LACZRKUWKHQVKS-UHFFFAOYSA-N
SMILES: NC1=CC(=C(OC2=CC=C(OC3=C(C=C(N)C=C3)C(F)(F)F)C=C2)C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results