accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 94598-32-0

IUPAC Name: 1-bromopentacosa-10,12-diyne
Synonyms: 1-bromopentacosa-10,12-diyne
Molecular Weight (g/mol): 423.52
Molecular Formula: C25H43Br
InChi Key: NUDHYRYEPCJLCL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#CC#CCCCCCCCCCBr
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results