accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 94624-12-1

IUPAC Name: pentan-1-ol
Synonyms: amyl alcohol alcool amylique n-C5H11OH pentylalkohol n-amylalkohol amyl alcohol, normal primary amyl alcohol amylalkohol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: AMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILES: CCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results