accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONH2

CAS RN 946409-08-1

IUPAC Name: 1-(4-methyl-3,4-dihydro-2H-1,4-benzoxazin-7-yl)methanamine
Synonyms: 1-(4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl)methanamine
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: PGIOCCIKSFJJMR-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1C=CC(CN)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results