accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClOFFF

CAS RN 94695-48-4

IUPAC Name: 2,3,4,5-tetrafluorobenzoyl chloride
Synonyms: 2,3,4,5-tetrafluorobenzoyl chloride
Molecular Weight (g/mol): 212.53
Molecular Formula: C7HClF4O
InChi Key: XWCKIXLTBNGIHV-UHFFFAOYSA-N
SMILES: FC1=CC(C(Cl)=O)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results