accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94695-50-8

IUPAC Name: ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate
Synonyms: ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate
Molecular Weight (g/mol): 264.18
Molecular Formula: C11H8F4O3
InChi Key: KWDVJYLIAJHEOW-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results