accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOO

CAS RN 947-62-6

IUPAC Name: 2-methoxynaphthalene-1-carboxylic acid
Synonyms: 2-methoxynaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: OSTYZAHQVPMQHI-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1C(O)=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)