accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94741-70-5

IUPAC Name: 4-amino-2-bromopyrimidine-5-carbonitrile
Synonyms: 4-amino-2-bromopyrimidine-5-carbonitrile
Molecular Weight (g/mol): 199.01
Molecular Formula: C5H3BrN4
InChi Key: CXYLLFGNJJCGHM-UHFFFAOYSA-N
SMILES: NC1=NC(Br)=NC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results