accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONN

CAS RN 948027-43-8

IUPAC Name: 5-(pyridin-3-yl)-7,11-diazatricyclo[7.3.1.0²,⁷]trideca-2,4-dien-6-one
Synonyms: 5-(pyridin-3-yl)-7,11-diazatricyclo[7.3.1.0²,⁷]trideca-2,4-dien-6-one
Molecular Weight (g/mol): 267.33
Molecular Formula: C16H17N3O
InChi Key: GOYASQWDLRLAPK-UHFFFAOYNA-N
SMILES: O=C1N2CC3CNCC(C3)C2=CC=C1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results