accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNH2

CAS RN 94838-59-2

IUPAC Name: tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate
Synonyms: tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate
Molecular Weight (g/mol): 236.32
Molecular Formula: C13H20N2O2
InChi Key: HOPALBZGTWDOTL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC1=CC=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)