accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94839-07-3

IUPAC Name: (2H-1,3-benzodioxol-5-yl)boronic acid
Synonyms: 2H-1,3-benzodioxol-5-ylboronic acid
Molecular Weight (g/mol): 165.94
Molecular Formula: C7H7BO4
InChi Key: CMHPUBKZZPSUIQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)