accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 949-42-8

CH3NNNClClClClClCl

CAS RN 949-42-8

IUPAC Name: 2-methyl-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-methyl-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 329.81
Molecular Formula: C6H3Cl6N3
InChi Key: LETDRANQSOEVCX-UHFFFAOYSA-N
SMILES: CC1=NC(=NC(=N1)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)