accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-11-4

N

CAS RN 95-11-4

IUPAC Name: bicyclo[2.2.1]hept-5-ene-2-carbonitrile
Synonyms: 5-norbornene-2-carbonitrile
Molecular Weight (g/mol): 119.17
Molecular Formula: C8H9N
InChi Key: BMAXQTDMWYDIJX-UHFFFAOYNA-N
SMILES: N#CC1CC2CC1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)