CAS RN 95-17-0
CAS RN 95-17-0
IUPAC Name:
bicyclo[2.2.1]hept-5-ene-2-carboxamide
Synonyms:
bicyclo[2.2.1]hept-5-ene-2-carboxamide
Molecular Weight (g/mol):
137.18
Molecular Formula:
C8H11NO
InChi Key:
ZTUUVDYQBLRAAC-UHFFFAOYNA-N
SMILES:
NC(=O)C1CC2CC1C=C2
1
–
1
of
1
results
1
5-Norbornene-2-carboxamide (mixture of isomers) 98.0+%, TCI America™
CAS: 95-17-0 Molecular Formula: C8H11NO Molecular Weight (g/mol): 137.182 MDL Number: MFCD00187896 InChI Key: ZTUUVDYQBLRAAC-UHFFFAOYSA-N Synonym: Bicyclo[2.2.1]hept-5-ene-2-carboxamide PubChem CID: 96112 IUPAC Name: bicyclo[2.2.1]hept-2-ene-5-carboxamide SMILES: C1C2CC(C1C=C2)C(=O)N