accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-21-6

IUPAC Name: 2-methyl-1,3-benzoxazole
Synonyms: 2-methylbenzoxazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: DQSHFKPKFISSNM-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2O1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)