accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-30-7

IUPAC Name: 1-(1,3-benzothiazol-2-ylsulfanyl)-N,N-diethylmethanethioamide
Synonyms: 1-(1,3-benzothiazol-2-ylsulfanyl)-N,N-diethylmethanethioamide
Molecular Weight (g/mol): 282.44
Molecular Formula: C12H14N2S3
InChi Key: LFMQNMXVVXHZCC-UHFFFAOYSA-N
SMILES: CCN(CC)C(=S)SC1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results