accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2Cl

CAS RN 95-51-2

IUPAC Name: 2-chloroaniline
Synonyms: 2-chloroaniline
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: AKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (6)
  • (5)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)