accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3F

CAS RN 95-52-3

IUPAC Name: 1-fluoro-2-methylbenzene
Synonyms: O-fluorotoluene
Molecular Weight (g/mol): 110.13
Molecular Formula: C7H7F
InChi Key: MMZYCBHLNZVROM-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1F
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (4)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)