accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2H3C

CAS RN 95-68-1

IUPAC Name: 2,4-dimethylaniline
Synonyms: 2,4-dimethylaniline m-xylidine meta-xylidine ccris 254 m-4-xylidine asym-m-xylidene
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: CZZZABOKJQXEBO-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)