accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClCH3NH2H2N

CAS RN 95-70-5

IUPAC Name: dihydrogen 2-methylbenzene-1,4-diamine dichloride
Synonyms: 2,5-diaminotoluene
Molecular Weight (g/mol): 195.09
Molecular Formula: C7H12Cl2N2
InChi Key: VQUHVWVGRKTIBH-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC(N)=CC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results