accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClCl

CAS RN 95-75-0

IUPAC Name: 1,2-dichloro-4-methylbenzene
Synonyms: 3,4-dichlorotoluene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: WYUIWKFIFOJVKW-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)