accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-85-2

IUPAC Name: 2-amino-4-chlorophenol
Synonyms: phenol, 2-amino-4-chloro-
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: SWFNPENEBHAHEB-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results