accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-87-4

IUPAC Name: 2,5-dimethylphenol
Synonyms: 2,5-xylenol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: NKTOLZVEWDHZMU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (1)
  • (8)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)