accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3C

CAS RN 95-93-2

IUPAC Name: 1,2,4,5-tetramethylbenzene
Synonyms: durene 2,5-dimethyl- (7CI) p-xylene p-xylene, 2,5-dimethyl- (7CI)
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: SQNZJJAZBFDUTD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C=C1C
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (5)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)