accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClCl

CAS RN 95-95-4

IUPAC Name: 2,4,5-trichlorophenol
Synonyms: 2,4,5-trichlorophenol collunosol dowicide 2 C6H3cl3O ccris 718 preventol I rcra waste number U230
Molecular Weight (g/mol): 197.44
Molecular Formula: C6H3Cl3O
InChi Key: LHJGJYXLEPZJPM-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)