accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 950-35-6

IUPAC Name: dimethyl 4-nitrophenyl phosphate
Synonyms: methyl paraoxon
Molecular Weight (g/mol): 247.14
Molecular Formula: C8H10NO6P
InChi Key: BAFQDKPJKOLXFZ-UHFFFAOYSA-N
SMILES: COP(=O)(OC)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results