accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNOOHNH

CAS RN 95051-10-8

IUPAC Name: 4-{7,12,17-triphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3(24),4,6,8,10,12,14,16,18(21),19-undecaen-2-yl}benzoic acid
Synonyms: 4-{7,12,17-triphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3(24),4,6,8,10,12,14,16,18(21),19-undecaen-2-yl}benzoic acid
Molecular Weight (g/mol): 658.76
Molecular Formula: C45H30N4O2
InChi Key: VPIIZALKBPNPDZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=C2C=CC(=N2)C(=C2NC(C=C2)=C(C2=CC=C(N2)C(=C2C=CC1=N2)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results