accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 950603-27-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(morpholine-4-sulfonyl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(morpholine-4-sulfonyl)benzoate
Molecular Weight (g/mol): 437.34
Molecular Formula: C17H12F5NO5S
InChi Key: FDXYCKHOIXBDOA-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2S(=O)(=O)N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results