accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 950670-18-5

IUPAC Name: 3-bromo-5-fluoropyridine-2-carbonitrile
Synonyms: 3-bromo-5-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 201
Molecular Formula: C6H2BrFN2
InChi Key: OLOSREGULYFXDD-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(N=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results