accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOOH3C

CAS RN 95091-92-2

IUPAC Name: N-methoxy-N-methylfuran-2-carboxamide
Synonyms: N-methoxy-N-methylfuran-2-carboxamide
Molecular Weight (g/mol): 155.15
Molecular Formula: C7H9NO3
InChi Key: ZPMBEUDMVUSUOS-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results