accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 951-78-0

IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 2'-deoxyuridine|deoxyuridine
Molecular Weight (g/mol): 228.2
Molecular Formula: C9H12N2O5
InChi Key: MXHRCPNRJAMMIM-SHYZEUOFSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)