accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNH2

CAS RN 951-87-1

IUPAC Name: 1,2-diphenylethane-1,2-diamine
Synonyms: 1,2-diphenylethane-1,2-diamine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: PONXTPCRRASWKW-UHFFFAOYNA-N
SMILES: NC(C(N)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results