accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 95162-14-4

IUPAC Name: 4-bromo-1-(triphenylmethyl)-1H-pyrazole
Synonyms: 4-bromo-1-(triphenylmethyl)pyrazole
Molecular Weight (g/mol): 389.3
Molecular Formula: C22H17BrN2
InChi Key: CPENTLJGGGSVAJ-UHFFFAOYSA-N
SMILES: BrC1=CN(N=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results