CAS RN 95197-95-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
5(6)-Tetramethylrhodamine isothiocyanate
5(6)-Tetramethylrhodamine isothiocyanate
CAS: 95197-95-8 Molecular Formula: C25H21N3O3S MDL Number: MFCD00063517 Synonym: TRITC; MRITC