accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95233-18-4

IUPAC Name: 3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
Synonyms: atovaquone
Molecular Weight (g/mol): 366.84
Molecular Formula: C22H19ClO3
InChi Key: BSJMWHQBCZFXBR-UHFFFAOYSA-N
SMILES: OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)