accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95233-30-0

IUPAC Name: 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 196.17
Molecular Formula: C8H11F3O2
InChi Key: LMEAZIIFLVDISW-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CC1)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)