accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNNHOHO

CAS RN 952514-79-3

IUPAC Name: [4-(1-phenyl-1H-1,3-benzodiazol-2-yl)phenyl]boronic acid
Synonyms: 4-(1-phenyl-1,3-benzodiazol-2-yl)phenylboronic acid
Molecular Weight (g/mol): 314.15
Molecular Formula: C19H15BN2O2
InChi Key: PBSIVXAPTBHFFV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results