accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 953-26-4

IUPAC Name: ethyl (2E)-3-(4-nitrophenyl)prop-2-enoate
Synonyms: ethyl (2E)-3-(4-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 221.21
Molecular Formula: C11H11NO4
InChi Key: PFBQVGXIMLXCQB-VMPITWQZSA-N
SMILES: CCOC(=O)\C=C\C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results